The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034147.1 | NP_001029319.1 | 6 - 7 | 666 | 115 |
GenBank mRNA | BC100091.1 | - | 6 - 7 | 666 | 115 |
FQ213073.1 | - | 6 - 7 | 712 | 115 |