The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037369.1 | NP_001032446.1 | 3 - 4 | 704 | 67 |
XM_006256599.2 | XP_006256661.1 | 2 - 3 | 944 | 67 | |
GenBank mRNA | AY787642.1 | - | 1 - 2 | 671 | 67 |
BC105761.1 | - | 3 - 4 | 704 | 67 | |
FQ218039.1 | - | 3 - 4 | 606 | 67 |