Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lace1
Entrez Gene ID
502479
Gene Name
lactation elevated 1
Gene Aliases
-
Chromosome Location
Chr.20: 47038824 - 47208812 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037656.1 NP_001032745.1 7 - 8 1155 116
GenBank mRNA BC107937.1 - 7 - 8 1155 116

View Full Product Details