The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013123.3 | NP_037255.3 | 7 - 8 | 891 | 125 |
XM_006244746.2 | XP_006244808.1 | 8 - 9 | 3000 | 125 | |
XM_006244747.2 | XP_006244809.1 | 8 - 9 | 1257 | 125 | |
XM_006244748.2 | XP_006244810.1 | 7 - 8 | 1161 | 125 | |
XM_006244749.2 | XP_006244811.1 | 9 - 10 | 1080 | 125 | |
XM_006244750.2 | XP_006244812.1 | 9 - 10 | 3072 | 125 | |
XM_006244751.2 | XP_006244813.1 | 8 - 9 | 991 | 125 | |
XM_006244752.2 | XP_006244814.1 | 8 - 9 | 1233 | 125 | |
XM_006244753.2 | XP_006244815.1 | 8 - 9 | 2352 | 125 | |
XM_006244754.2 | XP_006244816.1 | 7 - 8 | 2904 | 125 | |
XM_008766993.1 | XP_008765215.1 | 8 - 9 | 1008 | 125 | |
GenBank mRNA | AB079118.1 | - | 5 - 6 | 585 | 125 |
AB112436.1 | - | 9 - 10 | 1025 | 125 | |
AB112437.1 | - | 7 - 8 | 891 | 125 | |
AB112438.1 | - | 8 - 9 | 963 | 125 | |
M95578.1 | - | 8 - 9 | 1212 | 125 |