Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asnsd1
Entrez Gene ID
299507
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
RGD1306332
Chromosome Location
Chr.9: 52988042 - 53009054 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106763.1 NP_001100233.1 5 - 6 1999 109

View Full Product Details