Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Khdrbs2
Entrez Gene ID
170843
Gene Name
KH domain containing, RNA binding, signal transduction associated 2
Gene Aliases
Slm1
Chromosome Location
Chr.9: 39501431 - 40008702 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766952.1 XP_008765174.1 16 - 17 2240 131

View Full Product Details