The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080397.1 | NP_536322.1 | 5 - 6 | 792 | 89 |
XM_006244730.2 | XP_006244792.1 | 6 - 7 | 941 | 89 | |
XM_006244732.2 | XP_006244794.1 | 5 - 6 | 599 | 89 | |
XM_008766985.1 | XP_008765207.1 | 5 - 6 | 723 | 89 | |
XM_008766987.1 | XP_008765209.1 | 5 - 6 | 693 | 89 | |
GenBank mRNA | AF022729.1 | - | 5 - 6 | 792 | 89 |