The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022229.2 | NP_071565.2 | 11 - 12 | 1634 | 137 |
XM_006244932.2 | XP_006244994.1 | 11 - 12 | 1670 | 137 | |
GenBank mRNA | BC086507.1 | - | 11 - 12 | 1634 | 137 |
FQ228527.1 | - | 1 - 2 | 176 | 137 | |
X53585.1 | - | 10 - 11 | 1498 | 137 | |
X54793.1 | - | 10 - 11 | 1582 | 137 |