Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lix1
Entrez Gene ID
292381
Gene Name
Lix1 homolog (chicken)
Gene Aliases
-
Chromosome Location
Chr.1: 59156021 - 59212878 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106214.1 NP_001099684.1 1 - 2 515 71

View Full Product Details