The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017445.1 | NP_001017445.1 | 1 - 2 | 71 | 64 |
XM_006253617.2 | XP_006253679.1 | 1 - 2 | 114 | 64 | |
XM_008771488.1 | XP_008769710.1 | 1 - 2 | 114 | 64 | |
GenBank mRNA | BC079356.1 | - | 1 - 2 | 88 | 64 |
BC091224.1 | - | 1 - 2 | 71 | 64 |