The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106102.1 | NP_001099572.1 | 1 - 2 | 191 | 89 |
GenBank mRNA | BC166544.1 | - | 1 - 2 | 161 | 89 |