Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gkap1
Entrez Gene ID
361202
Gene Name
G kinase anchoring protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 6785428 - 6825145 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012160.1 NP_001012160.1 10 - 11 1185 74
XM_006253559.2 XP_006253621.1 10 - 11 1040 74
XM_006253561.2 XP_006253623.1 9 - 10 962 74
GenBank mRNA BC083717.1 - 10 - 11 1185 74

View Full Product Details