Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fastk
Entrez Gene ID
296741
Gene Name
Fas-activated serine/threonine kinase
Gene Aliases
-
Chromosome Location
Chr.4: 7260529 - 7264590 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006235892.2 XP_006235954.1 3 - 4 779 125
XM_006235894.2 XP_006235956.1 3 - 4 766 125
XM_006235895.1 XP_006235957.1 3 - 4 763 125
XM_006235896.2 XP_006235958.1 3 - 4 780 125
GenBank mRNA BC083715.1 - 3 - 4 783 125
FQ215763.1 - 4 - 5 641 125
FQ228972.1 - 3 - 4 702 125

View Full Product Details