Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rsbn1l
Entrez Gene ID
311987
Gene Name
round spermatid basic protein 1-like
Gene Aliases
RGD1308748
Chromosome Location
Chr.4: 10748745 - 10809142 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135872.1 NP_001129344.1 7 - 8 2308 140

View Full Product Details