Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pi15
Entrez Gene ID
301489
Gene Name
peptidase inhibitor 15
Gene Aliases
-
Chromosome Location
Chr.5: 791137 - 819233 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106917.1 NP_001100387.1 3 - 4 473 132

View Full Product Details