Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vcpip1
Entrez Gene ID
286761
Gene Name
valosin containing protein (p97)/p47 complex interacting protein 1
Gene Aliases
Vcip135
Chromosome Location
Chr.5: 9231180 - 9257821 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176857.3 NP_789827.3 1 - 2 2821 102
GenBank mRNA AB045378.2 - 1 - 2 2821 102
AF289091.1 - - 2879 102

View Full Product Details