The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025695.1 | NP_001020866.1 | 7 - 8 | 1034 | 151 |
XM_006237758.2 | XP_006237820.1 | 7 - 8 | 974 | 151 | |
GenBank mRNA | BC098736.1 | - | 7 - 8 | 1034 | 151 |
FQ215612.1 | - | 7 - 8 | 957 | 151 | |
FQ215908.1 | - | 7 - 8 | 957 | 151 | |
FQ227775.1 | - | 7 - 8 | 958 | 151 | |
FQ232542.1 | - | 7 - 8 | 957 | 151 |