Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gatsl3
Entrez Gene ID
360969
Gene Name
GATS protein-like 3
Gene Aliases
RGD1304774
Chromosome Location
Chr.14: 84447853 - 84457396 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025128.1 NP_001020299.1 3 - 4 423 118
XM_006251335.2 XP_006251397.1 3 - 4 491 118
XM_006251336.2 XP_006251398.1 3 - 4 491 118
GenBank mRNA BC091274.1 - 3 - 4 423 118

View Full Product Details