Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sf3a1
Entrez Gene ID
305479
Gene Name
splicing factor 3a, subunit 1
Gene Aliases
-
Chromosome Location
Chr.14: 84393182 - 84413931 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107235.1 NP_001100705.1 4 - 5 773 96

View Full Product Details