Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sec14l3
Entrez Gene ID
64543
Gene Name
SEC14-like 3 (S. cerevisiae)
Gene Aliases
Spf2
Chromosome Location
Chr.14: 84306466 - 84317241 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022608.1 NP_072130.1 9 - 10 799 80
GenBank mRNA AJ132352.1 - 9 - 10 814 80
BC162038.1 - 9 - 10 799 80

View Full Product Details