The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107231.2 | NP_001100701.1 | 1 - 2 | 1272 | 63 |
NM_001277214.1 | NP_001264143.1 | 1 - 2 | 1272 | 63 | |
NM_001277215.1 | NP_001264144.1 | 1 - 2 | 1272 | 63 | |
XM_006251292.2 | XP_006251354.1 | 1 - 2 | 1294 | 63 | |
GenBank mRNA | BC098801.1 | - | 1 - 2 | 151 | 63 |
CK479676.1 | - | 1 - 2 | 78 | 63 | |
EV764152.1 | - | 1 - 2 | 104 | 63 |