Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Camk2b
Entrez Gene ID
24245
Gene Name
calcium/calmodulin-dependent protein kinase II beta
Gene Aliases
Ck2b
Chromosome Location
Chr.14: 86208876 - 86297751 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042354.1 NP_001035813.1 16 - 17 1321 111
XM_006251170.2 XP_006251232.1 19 - 20 1651 111
XM_006251171.2 XP_006251233.1 18 - 19 1606 111
XM_006251172.2 XP_006251234.1 18 - 19 1579 111
XM_006251174.2 XP_006251236.1 17 - 18 1534 111
GenBank mRNA AF069731.1 - 2 - 3 243 111
X83375.1 - 16 - 17 1283 111

View Full Product Details