Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rasl10a
Entrez Gene ID
364190
Gene Name
RAS-like, family 10, member A
Gene Aliases
RGD1306100
Chromosome Location
Chr.14: 85311833 - 85313211 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108862.1 NP_001102332.1 2 - 3 387 167

View Full Product Details