The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012151.1 | NP_001012151.1 | 5 - 6 | 511 | 148 |
GenBank mRNA | BC079183.1 | - | 5 - 6 | 511 | 148 |