Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anxa3
Entrez Gene ID
25291
Gene Name
annexin A3
Gene Aliases
Anx3, LC3, LRRGT00047
Chromosome Location
Chr.14: 14371921 - 14426507 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012823.2 NP_036955.2 3 - 4 244 69
XM_006250694.2 XP_006250756.1 3 - 4 608 69
XM_006250696.2 XP_006250758.1 4 - 5 364 69
GenBank mRNA BC081856.1 - 2 - 3 156 69
FQ232970.1 - 3 - 4 235 69
M20559.1 - 3 - 4 192 69

View Full Product Details