Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stfa2l3
Entrez Gene ID
498087
Gene Name
stefin A2-like 3
Gene Aliases
RGD1560676
Chromosome Location
Chr.11: 67451585 - 67454007 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248415.2 XP_006248477.1 1 - 2 153 174

View Full Product Details