Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc58
Entrez Gene ID
288065
Gene Name
coiled-coil domain containing 58
Gene Aliases
RGD1564582
Chromosome Location
Chr.11: 67598419 - 67618704 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105875.1 NP_001099345.1 3 - 4 871 140

View Full Product Details