The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212519.2 | NP_997684.1 | 7 - 8 | 1372 | 95 |
XM_008768643.1 | XP_008766865.1 | 7 - 8 | 1666 | 95 | |
XM_008768644.1 | XP_008766866.1 | 7 - 8 | 1672 | 95 | |
XM_008768645.1 | XP_008766867.1 | 7 - 8 | 1670 | 95 | |
GenBank mRNA | AB162856.1 | - | 7 - 8 | 1235 | 95 |
BC098640.1 | - | 7 - 8 | 1372 | 95 | |
FQ228582.1 | - | - | 1417 | 95 |