Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exosc1
Entrez Gene ID
679140
Gene Name
exosome component 1
Gene Aliases
-
Chromosome Location
Chr.1: 261169798 - 261179865 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749119.3 XP_003749167.1 5 - 6 421 66

View Full Product Details