Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bloc1s2
Entrez Gene ID
293938
Gene Name
biogenesis of lysosomal organelles complex-1, subunit 2
Gene Aliases
RSEP1, Sep1
Chromosome Location
Chr.1: 263913901 - 263920980 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037349.2 NP_001032426.1 1 - 2 78 72
XM_008760401.1 XP_008758623.1 1 - 2 102 72
GenBank mRNA AY786315.1 - 1 - 2 59 72
CK220925.1 - 1 - 2 78 72
EF061226.2 - 1 - 2 56 72

View Full Product Details