Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spg21
Entrez Gene ID
300791
Gene Name
spastic paraplegia 21 homolog (human)
Gene Aliases
-
Chromosome Location
Chr.8: 70994532 - 71022302 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006987.1 NP_001006988.1 1 - 2 194 75
XM_006243251.2 XP_006243313.1 1 - 2 225 75
GenBank mRNA BC083760.1 - 1 - 2 194 75

View Full Product Details