Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gadl1
Entrez Gene ID
367181
Gene Name
glutamate decarboxylase-like 1
Gene Aliases
RGD1565145
Chromosome Location
Chr.8: 123989821 - 124234845 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766660.1 XP_008764882.1 5 - 6 719 102

View Full Product Details