Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bsn
Entrez Gene ID
29138
Gene Name
bassoon (presynaptic cytomatrix protein)
Gene Aliases
Znf231
Chromosome Location
Chr.8: 116873721 - 116965396 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019146.2 NP_062019.2 11 - 12 12013 143

View Full Product Details