The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007754.1 | NP_001007755.1 | 3 - 4 | 581 | 69 |
NM_001037555.1 | NP_001032644.1 | 4 - 5 | 773 | 69 | |
GenBank mRNA | AB202124.1 | - | 4 - 5 | 773 | 69 |
BC079237.1 | - | 3 - 4 | 581 | 69 |