Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rwdd2a
Entrez Gene ID
363110
Gene Name
RWD domain containing 2A
Gene Aliases
Rwdd2
Chromosome Location
Chr.8: 94243324 - 94246496 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108773.1 NP_001102243.1 1 - 2 233 149

View Full Product Details