Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rwdd2a
Entrez Gene ID
363110
Gene Name
RWD domain containing 2A
Gene Aliases
Rwdd2
Chromosome Location
Chr.8: 94243324 - 94246496 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243483.2 XP_006243545.1 1 - 2 133 67

View Full Product Details