Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Me1
Entrez Gene ID
24552
Gene Name
malic enzyme 1, NADP(+)-dependent, cytosolic
Gene Aliases
MOD1
Chromosome Location
Chr.8: 94256830 - 94368834 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012600.2 NP_036732.2 2 - 3 211 131
XM_008766439.1 XP_008764661.1 2 - 3 508 131
GenBank mRNA FQ228526.1 - 2 - 3 242 131

View Full Product Details