The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005548.1 | NP_001005548.1 | 11 - 12 | 1939 | 83 |
GenBank mRNA | BC082098.1 | - | 11 - 12 | 1939 | 83 |
FQ229860.1 | - | 11 - 12 | 1939 | 83 |