Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
P4htm
Entrez Gene ID
301008
Gene Name
prolyl 4-hydroxylase, transmembrane
Gene Aliases
Ph-4, RGD1311848
Chromosome Location
Chr.8: 117364895 - 117383381 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_217285.8 XP_217285.6 2 - 3 1101 65

View Full Product Details