Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P4htm
Entrez Gene ID
301008
Gene Name
prolyl 4-hydroxylase, transmembrane
Gene Aliases
Ph-4, RGD1311848
Chromosome Location
Chr.8: 117364895 - 117383381 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_217285.8 XP_217285.6 6 - 7 1744 77

View Full Product Details