Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss45
Entrez Gene ID
408244
Gene Name
protease, serine, 45
Gene Aliases
Tessp5
Chromosome Location
Chr.8: 119112726 - 119118264 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008864.1 NP_001008864.1 5 - 6 719 70

View Full Product Details