Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmod2
Entrez Gene ID
58814
Gene Name
tropomodulin 2
Gene Aliases
-
Chromosome Location
Chr.8: 82505947 - 82547569 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031613.1 NP_113801.1 5 - 6 681 71
XM_006243407.2 XP_006243469.1 6 - 7 1141 71
XM_006243408.2 XP_006243470.1 6 - 7 4343 71
XM_008766331.1 XP_008764553.1 6 - 7 733 71
GenBank mRNA U59240.1 - 5 - 6 681 71

View Full Product Details