Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmod3
Entrez Gene ID
300838
Gene Name
tropomodulin 3
Gene Aliases
-
Chromosome Location
Chr.8: 82432165 - 82492243 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011997.1 NP_001011997.1 7 - 8 807 107
GenBank mRNA BC079290.1 - 7 - 8 807 107
FQ229312.1 - 2 - 3 241 107

View Full Product Details