Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mei4
Entrez Gene ID
679805
Gene Name
meiosis-specific 4 homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.8: 89369417 - 89556942 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109378.1 NP_001102848.1 3 - 4 929 69

View Full Product Details