Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr82
Entrez Gene ID
686295
Gene Name
WD repeat domain 82
Gene Aliases
-
Chromosome Location
Chr.8: 114866769 - 114886208 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271306.1 NP_001258235.1 - 994 69

View Full Product Details