The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014004.1 | NP_001014026.1 | 6 - 7 | 1071 | 92 |
GenBank mRNA | BC083803.1 | - | 6 - 7 | 1071 | 92 |
FQ219637.1 | - | 1 - 2 | 133 | 92 |