Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdhr4
Entrez Gene ID
363144
Gene Name
cadherin-related family member 4
Gene Aliases
Cdh29, RGD1306603
Chromosome Location
Chr.8: 116771378 - 116779242 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243872.2 XP_006243934.1 17 - 18 2461 71

View Full Product Details