Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ip6k2
Entrez Gene ID
59268
Gene Name
inositol hexakisphosphate kinase 2
Gene Aliases
Ihpk2, Pius
Chromosome Location
Chr.8: 117574104 - 117599849 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021660.2 NP_067692.2 4 - 5 680 78
XM_008766564.1 XP_008764786.1 3 - 4 646 78
XM_008766565.1 XP_008764787.1 2 - 3 356 78
XM_008766566.1 XP_008764788.1 4 - 5 715 78
GenBank mRNA BC083574.1 - 4 - 5 680 78

View Full Product Details