Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmgn3
Entrez Gene ID
113990
Gene Name
high mobility group nucleosomal binding domain 3
Gene Aliases
-
Chromosome Location
Chr.8: 90628203 - 90665059 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201353.1 NP_001188282.1 5 - 6 394 114

View Full Product Details