Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Irak1bp1
Entrez Gene ID
300862
Gene Name
interleukin-1 receptor-associated kinase 1 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 90343307 - 90360085 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277283.1 NP_001264212.1 2 - 3 419 123
GenBank mRNA BC159433.1 - 2 - 3 411 123
FQ215646.1 - 2 - 3 353 123

View Full Product Details